The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145878.1 | NP_665885.1 | 2 - 3 | 297 | 57 |
GenBank mRNA | DQ255913.1 | - | 1 - 2 | 155 | 57 |
FQ210725.1 | - | 2 - 3 | 307 | 57 | |
FQ228381.1 | - | 2 - 3 | 309 | 57 | |
S69874.1 | - | 2 - 3 | 299 | 57 | |
S83247.1 | - | 2 - 3 | 314 | 57 | |
U13253.1 | - | 2 - 3 | 297 | 57 |