The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177425.3 | NP_803174.2 | 4 - 5 | 459 | 70 |
GenBank mRNA | BC086356.1 | - | 4 - 5 | 473 | 70 |
U44948.1 | - | 4 - 5 | 464 | 70 |