The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181627.2 | NP_853658.1 | - | 570 | 96 |
GenBank mRNA | AY291062.1 | - | - | 570 | 96 |
AY291063.2 | - | - | 765 | 96 | |
AY291064.2 | - | - | 582 | 96 | |
AY291065.2 | - | - | 931 | 96 | |
AY919342.1 | - | - | 387 | 96 | |
AY919343.1 | - | - | 549 | 96 | |
FJ517605.1 | - | - | 442 | 96 | |
FJ517606.1 | - | - | 284 | 96 | |
FQ213378.1 | - | - | 587 | 96 | |
FQ213584.1 | - | - | 587 | 96 | |
FQ214037.1 | - | - | 588 | 96 | |
FQ214498.1 | - | - | 598 | 96 | |
FQ227907.1 | - | - | 578 | 96 | |
FQ232927.1 | - | - | 587 | 96 |