Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psme3
Entrez Gene ID
287716
Gene Name
proteasome (prosome, macropain) activator subunit 3
Gene Aliases
Ab2-371
Chromosome Location
Chr.10: 89236256 - 89244041 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011894.1 NP_001011894.1 2 - 3 275 152
GenBank mRNA BC089889.1 - 2 - 3 275 152

View Full Product Details