Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jmjd4
Entrez Gene ID
287359
Gene Name
jumonji domain containing 4
Gene Aliases
RGD1307186
Chromosome Location
Chr.10: 45798964 - 45804188 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105784.1 NP_001099254.1 5 - 6 979 64

View Full Product Details