Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx26b
Entrez Gene ID
681665
Gene Name
DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B
Gene Aliases
-
Chromosome Location
Chr.X: 158835423 - 158887055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773666.1 XP_008771888.1 1 - 2 504 78

View Full Product Details