Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amelx
Entrez Gene ID
29160
Gene Name
amelogenin, X-linked
Gene Aliases
Amel
Chromosome Location
Chr.X: 26439197 - 26450495 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271077.1 NP_001258006.1 5 - 6 648 71
NM_001271078.1 NP_001258007.1 5 - 6 285 71
NM_019154.1 NP_062027.1 5 - 6 639 71
GenBank mRNA U01245.2 - 4 - 5 607 71
U07054.1 - 4 - 5 226 71
U67130.1 - 5 - 6 639 71

View Full Product Details