Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Camk2b
Entrez Gene ID
24245
Gene Name
calcium/calmodulin-dependent protein kinase II beta
Gene Aliases
Ck2b
Chromosome Location
Chr.14: 86208876 - 86297751 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042354.1 NP_001035813.1 19 - 20 1624 67
NM_001042356.1 NP_001035815.1 17 - 18 1366 67
NM_021739.2 NP_068507.2 19 - 20 1483 67
XM_006251170.2 XP_006251232.1 22 - 23 1954 67
XM_006251171.2 XP_006251233.1 21 - 22 1909 67
XM_006251172.2 XP_006251234.1 21 - 22 1882 67
XM_006251174.2 XP_006251236.1 20 - 21 1837 67
XM_006251176.2 XP_006251238.1 18 - 19 1518 67
XM_006251177.2 XP_006251239.1 18 - 19 1490 67
GenBank mRNA CB692429.1 - 2 - 3 171 67
M16112.1 - 19 - 20 1463 67
X83375.1 - 19 - 20 1586 67

View Full Product Details