Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tet3
Entrez Gene ID
680576
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
-
Chromosome Location
Chr.4: 115060702 - 115160802 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001057850.5 XP_001057850.2 1 - 2 1832 60

View Full Product Details