Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bai2
Entrez Gene ID
313058
Gene Name
brain-specific angiogenesis inhibitor 2
Gene Aliases
-
Chromosome Location
Chr.5: 148168516 - 148231587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107914.1 NP_001101384.1 4 - 5 1240 76
XM_006238934.1 XP_006238996.1 5 - 6 1348 76
XM_006238935.2 XP_006238997.1 5 - 6 1711 76
XM_006238936.1 XP_006238998.1 5 - 6 1348 76
XM_006238937.1 XP_006238999.1 5 - 6 1348 76
XM_006238938.1 XP_006239000.1 4 - 5 1183 76
XM_008764141.1 XP_008762363.1 5 - 6 1348 76
XM_008764142.1 XP_008762364.1 4 - 5 1183 76

View Full Product Details