Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lypla2
Entrez Gene ID
83510
Gene Name
lysophospholipase II
Gene Aliases
-
Chromosome Location
Chr.5: 154315033 - 154319629 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031342.1 NP_112632.1 9 - 10 736 100
GenBank mRNA AB021645.1 - 9 - 10 736 100
BC070503.1 - 9 - 10 742 100
FQ219970.1 - 3 - 4 328 99

View Full Product Details