Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sobp
Entrez Gene ID
309860
Gene Name
sine oculis-binding protein homolog (Drosophila)
Gene Aliases
RGD1560479, Sobpl, jcx1
Chromosome Location
Chr.20: 47731713 - 47910442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104640.1 NP_001098110.1 3 - 4 419 78
XM_006256574.2 XP_006256636.1 3 - 4 486 78
GenBank mRNA DQ503410.1 - 3 - 4 419 78

View Full Product Details