Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmco2
Entrez Gene ID
100360795
Gene Name
transmembrane and coiled-coil domains 2
Gene Aliases
-
Chromosome Location
Chr.5: 140019871 - 140024464 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729525.4 XP_002729571.1 1 - 2 626 142

View Full Product Details