Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tex28
Entrez Gene ID
690282
Gene Name
testis expressed 28
Gene Aliases
-
Chromosome Location
Chr.X: 156543891 - 156569470 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191103.1 NP_001178032.1 - 871 115
XM_008773612.1 XP_008771834.1 - 976 115

View Full Product Details