Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcun1d4
Entrez Gene ID
360928
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 4
Gene Aliases
RGD1310422
Chromosome Location
Chr.14: 37210721 - 37284612 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108359.1 NP_001101829.1 3 - 4 221 67
XM_008770158.1 XP_008768380.1 3 - 4 232 67

View Full Product Details