Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb49
Entrez Gene ID
305428
Gene Name
zinc finger and BTB domain containing 49
Gene Aliases
Zfp509, Znf509
Chromosome Location
Chr.14: 77322014 - 77343736 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107223.1 NP_001100693.1 3 - 4 1347 55

View Full Product Details