Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pygo2
Entrez Gene ID
295251
Gene Name
pygopus family PHD finger 2
Gene Aliases
RGD1308535
Chromosome Location
Chr.2: 188757165 - 188762285 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106447.1 NP_001099917.1 1 - 2 158 58
GenBank mRNA BC169054.1 - 1 - 2 284 58
FQ228430.1 - 1 - 2 257 58

View Full Product Details