Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl15
Entrez Gene ID
314111
Gene Name
kelch-like family member 15
Gene Aliases
RGD1563101
Chromosome Location
Chr.X: 63343006 - 63394190 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108021.1 NP_001101491.1 1 - 2 90 89
XM_006257023.2 XP_006257085.1 1 - 2 490 89

View Full Product Details