Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdpr
Entrez Gene ID
307852
Gene Name
pyruvate dehydrogenase phosphatase regulatory subunit
Gene Aliases
RGD1308307
Chromosome Location
Chr.19: 43107852 - 43149242 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107430.1 NP_001100900.1 10 - 11 1517 99
XM_006255596.2 XP_006255658.2 12 - 13 1772 99
XM_008772514.1 XP_008770736.1 11 - 12 1688 99

View Full Product Details