Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctcf
Entrez Gene ID
83726
Gene Name
CCCTC-binding factor (zinc finger protein)
Gene Aliases
-
Chromosome Location
Chr.19: 37600377 - 37649674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031824.1 NP_114012.1 5 - 6 1250 107
XM_006255536.1 XP_006255598.1 4 - 5 1200 107
XM_008772536.1 XP_008770758.1 3 - 4 1217 107
XM_008772537.1 XP_008770759.1 3 - 4 1217 107
XM_008772538.1 XP_008770760.1 3 - 4 1217 107
XM_008772539.1 XP_008770761.1 3 - 4 1217 107
GenBank mRNA AF133731.2 - 5 - 6 1286 107

View Full Product Details