Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dennd5b
Entrez Gene ID
100365679
Gene Name
DENN/MADD domain containing 5B
Gene Aliases
-
Chromosome Location
Chr.4: 183476093 - 183604937 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763472.1 XP_008761694.1 13 - 14 2996 105

View Full Product Details