Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspb11
Entrez Gene ID
685284
Gene Name
heat shock protein family B (small), member 11
Gene Aliases
-
Chromosome Location
Chr.5: 126856215 - 126882913 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131002.1 NP_001124474.1 4 - 5 383 123
XM_006238524.2 XP_006238586.2 4 - 5 389 123
XM_006238525.1 XP_006238587.1 6 - 7 648 123
XM_006238526.2 XP_006238588.1 6 - 7 617 123
XM_006238527.2 XP_006238589.1 5 - 6 550 123
XM_006238530.2 XP_006238592.1 4 - 5 367 123
XM_008763898.1 XP_008762120.1 6 - 7 634 123
XM_008763899.1 XP_008762121.1 3 - 4 306 123
XM_008763900.1 XP_008762122.1 5 - 6 533 123
GenBank mRNA BC162011.1 - 4 - 5 383 123

View Full Product Details