The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108948.1 | NP_001102418.1 | 6 - 7 | 618 | 68 |
XM_006232890.2 | XP_006232952.1 | 10 - 11 | 997 | 68 | |
XM_006232891.2 | XP_006232953.1 | 9 - 10 | 922 | 68 | |
GenBank mRNA | BC168687.1 | - | - | 855 | 68 |