Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbms1
Entrez Gene ID
362138
Gene Name
RNA binding motif, single stranded interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 46802870 - 47025178 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012184.1 NP_001012184.1 6 - 7 1079 117
XM_006234234.2 XP_006234296.1 6 - 7 836 117
XM_006234235.2 XP_006234297.1 6 - 7 1129 117
XM_006234236.2 XP_006234298.1 6 - 7 928 117
XM_006234237.2 XP_006234299.1 6 - 7 1688 117
XM_006234238.2 XP_006234300.1 6 - 7 836 117
XM_006234239.2 XP_006234301.1 7 - 8 1326 117
XM_006234241.2 XP_006234303.1 6 - 7 926 117
XM_008761860.1 XP_008760082.1 6 - 7 1417 117
GenBank mRNA BC087094.1 - 6 - 7 1079 117

View Full Product Details