Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhbdl3
Entrez Gene ID
287556
Gene Name
rhomboid, veinlet-like 3 (Drosophila)
Gene Aliases
Rhbdl4
Chromosome Location
Chr.10: 67677009 - 67731670 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105819.1 NP_001099289.1 3 - 4 546 60
XM_006246980.2 XP_006247042.1 2 - 3 583 60
XM_006246981.1 XP_006247043.1 3 - 4 1272 60
XM_006246982.2 XP_006247044.1 3 - 4 608 60

View Full Product Details