Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brd1
Entrez Gene ID
315210
Gene Name
bromodomain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 129676119 - 129724303 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108103.1 NP_001101573.1 3 - 4 1578 63
XM_006242194.2 XP_006242256.1 3 - 4 1574 63
XM_006242196.2 XP_006242258.1 3 - 4 1574 63
XM_006242197.2 XP_006242259.1 3 - 4 1574 63

View Full Product Details