Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mkl1
Entrez Gene ID
315151
Gene Name
megakaryoblastic leukemia (translocation) 1
Gene Aliases
BSAC, FLMKL1, MELODY, MKL1met, Mrtf-a
Chromosome Location
Chr.7: 122233196 - 122403780 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750385.3 XP_003750433.1 12 - 13 8099 76
XM_006242130.2 XP_006242192.1 13 - 14 3156 76
XM_008765806.1 XP_008764028.1 12 - 13 3769 76
XM_235497.8 XP_235497.6 13 - 14 2830 76
GenBank mRNA AB588919.1 - 13 - 14 2717 76
AB588920.1 - 12 - 13 2572 76
AB588921.1 - 13 - 14 2699 76

View Full Product Details