The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107254.1 | NP_001100724.1 | 11 - 12 | 2172 | 65 |
GenBank mRNA | BC168894.1 | - | 11 - 12 | 2193 | 65 |