Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp10
Entrez Gene ID
63995
Gene Name
dual specificity phosphatase 10
Gene Aliases
Mkp-5, Mkp5
Chromosome Location
Chr.13: 104284660 - 104321455 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105734.1 NP_001099204.1 2 - 3 964 83

View Full Product Details