Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcl11b
Entrez Gene ID
314423
Gene Name
B-cell CLL/lymphoma 11B (zinc finger protein)
Gene Aliases
Ctip2
Chromosome Location
Chr.6: 131834097 - 131926438 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277287.1 NP_001264216.1 2 - 3 424 90
GenBank mRNA JF520429.1 - 2 - 3 424 90

View Full Product Details