Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ranbp9
Entrez Gene ID
364686
Gene Name
RAN binding protein 9
Gene Aliases
RGD1561940, RanBPM
Chromosome Location
Chr.17: 24020874 - 24103771 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771586.1 XP_008769808.1 2 - 3 650 67

View Full Product Details