The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134447.1 | NP_001127919.1 | 9 - 10 | 1148 | 69 |
GenBank mRNA | BC167106.1 | - | 9 - 10 | 1148 | 69 |