Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo15
Entrez Gene ID
361354
Gene Name
F-box protein 15
Gene Aliases
-
Chromosome Location
Chr.18: 81785408 - 81882796 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108436.1 NP_001101906.1 8 - 9 1198 150

View Full Product Details