The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022245.1 | NP_071581.1 | 4 - 5 | 386 | 192 |
GenBank mRNA | AF007107.1 | - | 4 - 5 | 407 | 250 |
AF007108.1 | - | 4 - 5 | 349 | 192 | |
BC086945.1 | - | 4 - 5 | 380 | 192 | |
D13205.1 | - | 4 - 5 | 386 | 192 | |
FQ209861.1 | - | 4 - 5 | 377 | 192 | |
FQ210790.1 | - | 4 - 5 | 374 | 192 | |
FQ224354.1 | - | 4 - 5 | 377 | 192 |