Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pqlc1
Entrez Gene ID
361352
Gene Name
PQ loop repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.18: 76767985 - 76805773 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013189.1 NP_001013207.1 2 - 3 463 70
XM_006254952.2 XP_006255014.1 3 - 4 755 70
XM_006254953.2 XP_006255015.1 3 - 4 1148 70
XM_008772155.1 XP_008770377.1 2 - 3 923 70
GenBank mRNA BC079072.1 - 2 - 3 921 70
BC088186.1 - 2 - 3 463 70

View Full Product Details