Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sbk2
Entrez Gene ID
691411
Gene Name
SH3 domain binding kinase family, member 2
Gene Aliases
RGD1563279
Chromosome Location
Chr.1: 72425728 - 72433494 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127539.2 NP_001121011.1 3 - 4 655 123
GenBank mRNA BC161893.1 - 3 - 4 655 123

View Full Product Details