Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssc5d
Entrez Gene ID
308341
Gene Name
scavenger receptor cysteine rich family, 5 domains
Gene Aliases
RGD1565772
Chromosome Location
Chr.1: 72443756 - 72462293 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134545.1 NP_001128017.1 4 - 5 604 93
XM_006228257.1 XP_006228319.1 4 - 5 1350 93

View Full Product Details