Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssc5d
Entrez Gene ID
308341
Gene Name
scavenger receptor cysteine rich family, 5 domains
Gene Aliases
RGD1565772
Chromosome Location
Chr.1: 72443756 - 72462293 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134545.1 NP_001128017.1 7 - 8 1332 76
XM_006228257.1 XP_006228319.1 7 - 8 2078 76

View Full Product Details