Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt1
Entrez Gene ID
171409
Gene Name
troponin T type 1 (skeletal, slow)
Gene Aliases
Fang2, sTnT, tnTs
Chromosome Location
Chr.1: 72889270 - 72899629 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277260.1 NP_001264189.1 7 - 8 244 92
NM_001277261.2 NP_001264190.1 6 - 7 211 92
NM_001277262.2 NP_001264191.1 6 - 7 214 92
NM_134388.2 NP_599215.2 7 - 8 247 92
GenBank mRNA AF399874.1 - 6 - 7 195 92
AY334079.1 - 6 - 7 191 92
AY334080.1 - 6 - 7 188 92
AY334081.1 - 5 - 6 158 92
AY334082.1 - 5 - 6 155 92

View Full Product Details