Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnnt1
Entrez Gene ID
171409
Gene Name
troponin T type 1 (skeletal, slow)
Gene Aliases
Fang2, sTnT, tnTs
Chromosome Location
Chr.1: 72889270 - 72899629 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277260.1 NP_001264189.1 1 - 2 46 69
NM_001277261.2 NP_001264190.1 1 - 2 46 69
NM_001277262.2 NP_001264191.1 1 - 2 46 69
NM_134388.2 NP_599215.2 1 - 2 46 69

View Full Product Details