Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hspbp1
Entrez Gene ID
246146
Gene Name
HSPA binding protein, cytoplasmic cochaperone 1
Gene Aliases
-
Chromosome Location
Chr.1: 72732418 - 72756087 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139261.1 NP_640354.1 3 - 4 535 118
XM_006228248.1 XP_006228310.1 3 - 4 762 118
XM_006228249.2 XP_006228311.1 3 - 4 527 118
XM_006228250.2 XP_006228312.1 3 - 4 591 118
GenBank mRNA AF187860.1 - 3 - 4 535 118
BC072541.1 - 3 - 4 571 118
FQ232199.1 - 3 - 4 591 118

View Full Product Details