Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem86b
Entrez Gene ID
690610
Gene Name
transmembrane protein 86B
Gene Aliases
-
Chromosome Location
Chr.1: 72785001 - 72787367 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109604.1 NP_001103074.1 1 - 2 141 98
GenBank mRNA FQ219460.1 - 1 - 2 55 98

View Full Product Details