The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106924.1 | NP_001100394.1 | 7 - 8 | 1131 | 73 |
XM_006245373.2 | XP_006245435.1 | 8 - 9 | 1280 | 73 | |
XM_006245374.2 | XP_006245436.1 | 8 - 9 | 1423 | 73 | |
XM_006245375.2 | XP_006245437.1 | 8 - 9 | 1182 | 73 | |
GenBank mRNA | FQ224987.1 | - | 1 - 2 | 107 | 73 |
FQ225203.1 | - | 1 - 2 | 60 | 73 | |
FQ225560.1 | - | 1 - 2 | 107 | 73 | |
FQ226825.1 | - | 3 - 4 | 273 | 73 | |
FQ227560.1 | - | 1 - 2 | 119 | 73 | |
FQ232493.1 | - | 1 - 2 | 119 | 73 | |
FQ232683.1 | - | 4 - 5 | 447 | 73 | |
FQ233663.1 | - | 3 - 4 | 272 | 73 | |
FQ234937.1 | - | 2 - 3 | 204 | 73 | |
FQ235215.1 | - | 1 - 2 | 79 | 73 |