Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfand2b
Entrez Gene ID
363253
Gene Name
zinc finger, AN1 type domain 2B
Gene Aliases
RGD1306260
Chromosome Location
Chr.9: 82370897 - 82373843 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025745.1 NP_001020916.1 1 - 2 252 132
GenBank mRNA BC099215.1 - 1 - 2 252 132

View Full Product Details