Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc23a3
Entrez Gene ID
367298
Gene Name
solute carrier family 23, member 3
Gene Aliases
-
Chromosome Location
Chr.9: 82328007 - 82338576 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109006.1 NP_001102476.1 8 - 9 1210 72
XM_006245260.2 XP_006245322.1 9 - 10 1496 72
XM_008767280.1 XP_008765502.1 8 - 9 1389 72
XM_008767281.1 XP_008765503.1 9 - 10 1217 72

View Full Product Details