The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024891.1 | NP_001020062.1 | 6 - 7 | 1113 | 61 |
XM_008767250.1 | XP_008765472.1 | 6 - 7 | 1753 | 61 | |
GenBank mRNA | BC097416.1 | - | 6 - 7 | 1113 | 61 |