Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajb2
Entrez Gene ID
689593
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 2
Gene Aliases
-
Chromosome Location
Chr.9: 82436450 - 82444669 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245276.1 XP_006245338.1 3 - 4 390 62
XM_006245278.2 XP_006245340.1 3 - 4 412 62
XM_006245279.2 XP_006245341.1 3 - 4 339 62
XM_006245280.1 XP_006245342.1 3 - 4 390 62
XM_006245283.1 XP_006245345.1 3 - 4 390 62
GenBank mRNA BC166710.1 - 3 - 4 327 62

View Full Product Details