Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpine2
Entrez Gene ID
29366
Gene Name
serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2
Gene Aliases
Gdnpn1, PI-7, Pn-1, Spin4
Chromosome Location
Chr.9: 85560852 - 85626125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019197.1 NP_062070.1 7 - 8 1196 64
XM_006245161.2 XP_006245223.1 7 - 8 1859 64
XM_006245162.2 XP_006245224.1 8 - 9 1323 64
XM_008767212.1 XP_008765434.1 7 - 8 1183 64
GenBank mRNA CF110312.1 - 3 - 4 354 64
DV726078.1 - 1 - 2 76 64
M17784.1 - 6 - 7 1072 64

View Full Product Details