Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alpp
Entrez Gene ID
100359675
Gene Name
alkaline phosphatase, placental
Gene Aliases
-
Chromosome Location
Chr.9: 94189961 - 94192960 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002730058.2 XP_002730104.2 7 - 8 1000 71

View Full Product Details