Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc16a14
Entrez Gene ID
316578
Gene Name
solute carrier family 16, member 14
Gene Aliases
-
Chromosome Location
Chr.9: 92503602 - 92531167 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108229.1 NP_001101699.1 4 - 5 1057 128

View Full Product Details